Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMHR2, Mouse, Clone: MIG7, Abnova™

Catalog No. 89111200 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-200 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-200 Supplier Abnova Corporation Supplier No. MAB3008

Mouse monoclonal antibody raised against synthetic peptide of AMHR2.

This gene encodes the receptor for the anti-Mullerian hormone (AMH) which, in addition to testosterone, results in male sex differentiation. AMH and testosterone are produced in the testes by different cells and have different effects. Testosterone promotes the development of male genitalia while the binding of AMH to the encoded receptor prevents the development of the mullerian ducts into uterus and Fallopian tubes. Mutations in this gene are associated with persistent Mullerian duct syndrome type II. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: NANYSHLPPPGSPG

Specifications

Antigen AMHR2
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone MIG7
Conjugate Unconjugated
Description anti-Mullerian hormone receptor, type II
Formulation In PBS, pH 7.4 (0.09% sodium azide)
Gene AMHR2
Gene Alias AMHR, MISR2, MISRII
Gene Symbols AMHR2
Host Species Mouse
Immunogen A synthetic peptide (conjugated with Hsp70) corresponding to amino acids 117-130 of human AMHR2.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269
Target Species Human, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.