Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amphetamine, Mouse, Clone: 3B71D5, Abnova™

Catalog No. 89134987 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-134-987 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-134-987 Supplier Abnova Corporation Supplier No. MAB1538
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against Amphetamine.

Specifications

Antigen Amphetamine
Applications ELISA
Classification Monoclonal
Clone 3B71D5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against Amphetamine.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM potassium phosphate buffer, 0.85% NaCl, pH 7.2 (0.05% sodium azide)
Host Species Mouse
Immunogen Amphetamine conjugated with BSA.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody react with Amphetamine and shows low cross-reactivity (less than 0.01%) against Methamphetamine.
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.