Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AMY2A/AMY2B Mouse anti-Human, Monoclonal Antibody, Abnova™

Catalog No. 89110902 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-902 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-902 Supplier Abnova Corporation Supplier No. MAB2216
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native AMY2A/AMY2B.

Specifications

Antigen AMY2A/AMY2B
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native AMY2A/AMY2B.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4
Gene Alias AMY2/AMY2B/PA
Gene Symbols AMY2A
Host Species Mouse
Immunogen Native purified AMY2A/AMY2B from human pancreas.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 279/280
Test Specificity These antibodies specifically recognize human pancreatic amylase.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.