Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid-beta 1-40, Mouse, Clone: BDI350, Abnova™

Catalog No. 89111651 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-651 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-651 Supplier Abnova Corporation Supplier No. MAB3913
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of Amyloid-beta 1-40.

This gene encodes a cell surface receptor and transmembrane precursor protein that is cleaved by secretases to form a number of peptides. Some of these peptides are secreted and can bind to the acetyltransferase complex APBB1/TIP60 to promote transcriptional activation, while others form the protein basis of the amyloid plaques found in the brains of patients with Alzheimer disease. Mutations in this gene have been implicated in autosomal dominant Alzheimer disease and cerebroarterial amyloidosis (cerebral amyloid angiopathy). Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen Amyloid-beta 1-40
Applications ELISA, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone BDI350
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Amyloid-beta 1-40.
Dilution ELISA (0.1 ug/mL) Western Blot (1 ug/mL HRP/ECL detection) Immunocytochemistry (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 2X PBS (PEG, sucrose, 0.09% sodium azide)
Gene APP
Gene Alias AAA/ABETA/ABPP/AD1/APPI/CTFgamma/CVAP/PN2
Gene Symbols APP
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human Amyloid-beta 1-40.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 351
Test Specificity Specifically reacts with the C-terminus of human beta Amyloid 1-40. Does not cross-react with beta Amyloid 1-42.
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.