Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid-beta, Mouse, Clone: 252Q6, Abnova™

Catalog No. 89110727 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-727 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-727 Supplier Abnova Corporation Supplier No. MAB1957

Mouse monoclonal antibody raised against synthetic peptide of Amyloid-beta.

Specifications

Antigen Amyloid-beta
Applications ELISA, Western Blot
Classification Monoclonal
Clone 252Q6
Conjugate Unconjugated
Description amyloid beta (A4) precursor protein
Formulation In PBS, pH 7.3 (0.08% sodium azide)
Gene App
Gene Alias AL024401, Abeta, Adap, Cvap, E030013M08Rik, appican, betaAPP
Gene Symbols App
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 1-20 of mouse Amyloid-beta.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11820
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.