Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Amyloid-beta, Mouse, Clone: OM16, Abnova™

Catalog No. 89135903 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-903 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-903 Supplier Abnova Corporation Supplier No. MAB5145

Mouse monoclonal antibody raised against synthetic peptide of Amyloid-beta.

Specifications

Antigen Amyloid-beta
Applications ELISA, Western Blot
Classification Monoclonal
Clone OM16
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of Amyloid-beta.
Dilution ELISA (1 ug/mL) Western Blot (10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 10mM sodium phosphate buffer, pH 7.4 (0.15M NaCl, 0.09% sodium azide)
Host Species Mouse
Immunogen A synthetic peptide corresponding to human Amyloid-beta.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts with beta-AP1-40 and secretory amyloid protein precursor (sAPP), but not with beta-AP17-42.
Target Species Human
Content And Storage Store at -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.