Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKS3, Mouse, Clone: 1B2, Abnova™

Catalog No. 89107587 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-587 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-587 Supplier Abnova Corporation Supplier No. H00124401M05
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ANKS3.

Sequence: MKDIQGWTALFHCTSAGHQHMVRFLLDSGANANVREPICGFTPLMEAAAAGHEIIVQYFLNHGVKVDARDHSGATARMLAKQYGHMKIVALMDTYSPSLPKSLYRSPEKYEDLSSSDESCPAPQRQRPCRKKGVSIHEGPRALARITGIGLGGRAPRPRYEQAPPRGYVTFNSSGENPLEEEGLCCRDVTSPINERDVESSSSSSSREEHAFCANLGPVQSSSSSEGLARAQGLSSEASVESNEDSDHACKSSARKQAKSYMKTKNPDSQWPPRTATDREGFLAESSPQTQRAPYSGPQDLAALLEQIGCLKYLQVFEEQDVDLRIFLTLTESDLKEIGITLFGPKRKMTSAIARWHSSARPPGDALELAYADRLEAEMQELAIQLHKRCEEVEATRGQVCQEQELRAVVESCLLEQDRAREDLQARLRETWALARDAALVLDQLRACQAELSSRVRQDQPPGAATLGLAVPPADSKGWQASLQAMSLPELSGALEDRVREMGQALCLVTQSLEKLQVLSGKKWRET

Specifications

Antigen ANKS3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1B2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant ANKS3.
Formulation PBS with no preservative; pH 7.4
Gene ANKS3
Gene Accession No. BC050387.1
Gene Alias FLJ32345/FLJ32767/KIAA1977
Gene Symbols ANKS3
Host Species Mouse
Immunogen ANKS3 (AAH50387.1,1 a.a. ∽ 527 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124401
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.