Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANKS4B, Mouse, Clone: 3E3, Abnova™

Catalog No. 89107738 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-738 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-738 Supplier Abnova Corporation Supplier No. H00257629M03
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant ANKS4B.

Sequence: MSTRYHQAASDSYLELLKEATKRDLNLSDEDGMTPTLLAAYHGNLEALEIICSRGGDPDRCDIWGNTPLHFAASNGHAHCVSFLVNFGANIFALDNDLQTPLDAAASREQNECVALLDKAATAQNIMNPKKVTRLKEQAQKNARRQIKECERLQEKHQNKMAHTYSKEESGTLSSSKGTFSRSSPSNASAPGTFGSLSKGIKDTFKIKFKKNKDTAEQVGKEGRSGQRNVMEVFREEEEDSFSGDFKEKLQLSAEEDGSVHHESILNRPGLGSIVFRRNRISSPEDISDSKREFGFKLPSELLQRQGASEADEGAADEEGEENGLKDDLPWDDDEVEWEEDVVDATPLEVFLLSQHLEEFLPIFKREQIDLEALLLCSDEDLQSIQMQLGPRKKVLNAINRRKQVLQQPGQLVDTSL

Specifications

Antigen ANKS4B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 3E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant ANKS4B.
Formulation PBS with no preservative; pH 7.4
Gene ANKS4B
Gene Accession No. NM_145865.1
Gene Alias FLJ38819/HARP/MGC133380/MGC133381
Gene Symbols ANKS4B
Host Species Mouse
Immunogen ANKS4B (NP_665872.1,1 a.a. ∽ 417 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Protein A Purification
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257629
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.