Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ANO1, Mouse, Clone: DG1/447, Abnova™

Catalog No. 89109413 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-109-413 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-109-413 Supplier Abnova Corporation Supplier No. MAB11243
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant ANO1.

Specifications

Antigen ANO1
Applications Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone DG1/447
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against partial recombinant ANO1.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.25-0.5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation Bioreactor concentrate (0.05% sodium azide)
Gene ANO1
Gene Alias FLJ10261/ORAOV2/TAOS2/TMEM16A
Gene Symbols ANO1
Host Species Mouse
Immunogen Recombinant protein corresponding to human ANO1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55107
Target Species Human
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.