Learn More
AP1G2, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant AP1G2.
Supplier: Abnova Corporation H00008906A01
Description
Adaptins are important components of clathrin-coated vesicles transporting ligand-receptor complexes from the plasma membrane or from the trans-Golgi network to lysosomes. The adaptin family of proteins is compsed of four classes of molecules named alpha, beta-, beta prime- and gamma- adaptins. Adaptins, together with medium and small subunits, form a heterotetrameric complex called an adaptor, whose role is to promote the formation of clathrin-coated pits and vesicles. The protein encoded by this gene is a gamma-adaptin protein and it belongs to the adaptor complexes large subunits family. This protein along with the complex is thought to function at some trafficking step in the complex pathways between the trans-Golgi network and the cell surface. There are two alternatively spliced transcript variants of this gene encoding the same protein. [provided by RefSeq
Sequence: RPPENPALLLITITATNFSEGDVTHFICQAAVPKSLQLQLQAPSGNTVPARGGLPITQLFRILNPNKAPLRLKLRLTYDHFHQSVQEIFEVNNLPVESWQSpecifications
AP1G2 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant AP1G2. | |
AP1G2 | |
G2AD | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_003917 | |
AP1G2 | |
AP1G2 (NP_003908, 686 a.a. to 785 a.a) partial recombinant protein with GST tag. | |
RUO | |
8906 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only