Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APAF1, Mouse, Clone: 2E10, Abnova™

Catalog No. 89111118 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-118 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-118 Supplier Abnova Corporation Supplier No. MAB2746
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of APAF1.

This gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Antigen APAF1
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 2E10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of APAF1.
Dilution Western Blot (0.25-0.5 mg/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.02% sodium azide)
Gene APAF1
Gene Alias CED4/DKFZp781B1145
Gene Symbols APAF1
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids near C-terminus of human APAF1.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.