Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APIP, Mouse, Clone: 19F461, Abnova™

Catalog No. 89107873 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-107-873 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-107-873 Supplier Abnova Corporation Supplier No. MAB0105
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant APIP.

APIP is an APAF1 (MIM 602233)-interacting protein that acts as a negative regulator of ischemic/hypoxic injury (Cho et al., 2004 [PubMed 15262985]).[supplied by OMIM

Specifications

Antigen APIP
Applications Western Blot
Classification Monoclonal
Clone 19F461
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant APIP.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% BSA, 0.05% sodium azide)
Gene APIP
Gene Alias APIP2/CGI-29/CGI29/MMRP19/dJ179L10.2
Gene Symbols APIP
Host Species Mouse
Immunogen Recombinant His fusion protein corresponding to full length human APIP.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51074
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.