Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOA5, Mouse, Clone: 4H8H8E2, Abnova™

Catalog No. 89110972 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-972 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-972 Supplier Abnova Corporation Supplier No. MAB2375
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against full-length recombinant APOA5.

The protein encoded by this gene is an apolipoprotein and an important determinant of plasma triglyceride levels, a major risk factor for coronary artery disease. It is a component of high density lipoprotein and is highly similar to a rat protein that is upregulated in response to liver injury. This gene uses alternate polyadenylation sites and is located proximal to the apolipoprotein gene cluster on chromosome 11q23. [provided by RefSeq

Specifications

Antigen APOA5
Applications ELISA
Classification Monoclonal
Clone 4H8H8E2
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against full length recombinant APOA5.
Dilution The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide)
Gene APOA5
Gene Alias APOA-V/APOAV/MGC126836/MGC126838/RAP3
Gene Symbols APOA5
Host Species Mouse
Immunogen Recombinant protein corresponding to full length human APOA5.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116519
Target Species Human
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.