Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOB, Mouse, Clone: 1605, Abnova™

Catalog No. 89111671 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-671 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-671 Supplier Abnova Corporation Supplier No. MAB4019
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native APOB.

This gene product is the main apolipoprotein of chylomicrons and low density lipoproteins. It occurs in plasma as two main isoforms, apoB-48 and apoB-100: the former is synthesized exclusively in the gut and the latter in the liver. The intestinal and the hepatic forms of apoB are encoded by a single gene from a single, very long mRNA. The two isoforms share a common N-terminal sequence. The shorter apoB-48 protein is produced after RNA editing of the apoB-100 transcript at residue 2180 (CAA->UAA), resulting in the creation of a stop codon, and early translation termination. Mutations in this gene or its regulatory region cause hypobetalipoproteinemia, normotriglyceridemic hypobetalipoproteinemia, and hypercholesterolemia due to ligand-defective apoB, diseases affecting plasma cholesterol and apoB levels. [provided by RefSeq

Specifications

Antigen APOB
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone 1605
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native APOB.
Dilution The optimal working dilution should be determined by the end user.
Formulation In 15mM potassium phosphate buffer, 150mM NaCl, pH 7.2 (0.09% sodium azide)
Gene APOB
Gene Alias FLDB
Gene Symbols APOB
Host Species Mouse
Immunogen Native purified APOB from human serum.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338
Test Specificity No cross reactivity with Apo A-I or Apo A-II.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.