Learn More
APOBEC3G, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant APOBEC3G.
Supplier: Abnova Corporation H00060489A01
Description
This gene is a member of the cytidine deaminase gene family. It is one of seven related genes or pseudogenes found in a cluster, thought to result from gene duplication, on chromosome 22. Members of the cluster encode proteins that are structurally and functionally related to the C to U RNA-editing cytidine deaminase APOBEC1. It is thought that the proteins may be RNA editing enzymes and have roles in growth or cell cycle control. The protein encoded by this gene has been found to be a specific inhibitor of human immunodeficiency virus-1 (HIV-1) infectivity. [provided by RefSeq
Sequence: LHRDQEYEVTWYISWSPCTKCTRDMATFLAEDPKVTLTIFVARLYYFWDPDYQEALRSLCQKRDGPRATMKIMNYDEFQHCWSKFVYSQRELFEPWNNLPKYSpecifications
APOBEC3G | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant APOBEC3G. | |
APOBEC3G | |
ARP9/CEM15/FLJ12740/MDS019/bK150C2.7/dJ494G10.1 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
ELISA, Western Blot | |
Unconjugated | |
50% glycerol | |
NM_021822 | |
APOBEC3G | |
APOBEC3G (NP_068594, 80 a.a. to 181 a.a) partial recombinant protein with GST tag. | |
RUO | |
60489 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only