Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APOM, Mouse, Clone: 8F12C6B8, Abnova™

Catalog No. 89108631 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-631 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-631 Supplier Abnova Corporation Supplier No. MAB10445
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant APOM.

The protein encoded by this gene is an apolipoprotein and member of the lipocalin protein family. It is found associated with high density lipoproteins and to a lesser extent with low density lipoproteins and triglyceride-rich lipoproteins. The encoded protein is secreted through the plasma membrane but remains membrane-bound, where it is involved in lipid transport. Two transcript variants encoding two different isoforms have been found for this gene, but only one of them has been fully characterized. [provided by RefSeq

Specifications

Antigen APOM
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 8F12C6B8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant APOM.
Dilution Western Blot (1:500-1:2000) Immunofluorescence (1:200-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.03% sodium azide)
Gene APOM
Gene Alias G3a/HSPC336/MGC22400/NG20
Gene Symbols APOM
Host Species Mouse
Immunogen Recombinant protein corresponding to human APOM.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55937
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.