Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AR, Mouse, Clone: 2F12, Abnova™

Catalog No. 89113690 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
3 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-690 3 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-690 Supplier Abnova Corporation Supplier No. MAB9566

Mouse monoclonal antibody raised against recombinant AR.

The androgen receptor gene is more than 90kb long and codes for a protein that has 3 major functional domains: the N-terminal domain, DNA-binding domain, and androgen-binding domain. The protein functions as a steroid-hormone activated transcription factor. Upon binding the hormone ligand, the receptor dissociates from accessory proteins, translocates into the nucleus, dimerizes, and then stimulates transcription of androgen responsive genes. This gene contains 2 polymorphic trinucleotide repeat segments that encode polyglutamine and polyglycine tracts in the N-terminal transactivation domain of its protein. Expansion of the polyglutamine tract causes spinal bulbar muscular atrophy (Kennedy disease). Mutations in this gene are also associated with complete androgen insensitivity (CAIS). Two alternatively spliced variants encoding distinct isoforms have been described. [provided by RefSeq

Specifications

Antigen AR
Applications Immunohistochemistry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2F12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant AR.
Dilution Immunohistochemistry (1:25) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from tissue culture supernatant (0.09% sodium azide)
Gene AR
Gene Alias AIS/DHTR/HUMARA/KD/NR3C4/SBMA/SMAX1/TFM
Gene Symbols AR
Host Species Mouse
Immunogen Recombinant protein corresponding to 321 amino acids near N-terminus of human AR.
Quantity 3 x 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 367
Target Species Human
Content And Storage Store at 4°C on dry atmosphere. After reconstitution with sterile distilled water store at 4°C for at least two months. For long term storage store at -20°C or below. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.