Learn More
ARF1, Mouse, Polyclonal Antibody, Abnova™
Mouse polyclonal antibody raised against a partial recombinant ARF1.
Supplier: Abnova Corporation H00000375A02
Description
ADP-ribosylation factor 1 (ARF1) is a member of the human ARF gene family. The family members encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking as activators of phospholipase D. The gene products, including 6 ARF proteins and 11 ARF-like proteins, constitute a family of the RAS superfamily. The ARF proteins are categorized as class I (ARF1, ARF2 and ARF3), class II (ARF4 and ARF5) and class III (ARF6), and members of each class share a common gene organization. The ARF1 protein is localized to the Golgi apparatus and has a central role in intra-Golgi transport. Multiple alternatively spliced transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: FQNTQGLIFVVDSNDRERVNEAREELMRMLAEDELRDAVLLVFANKQDLPNAMNAAEITDKLGLHSLRHRNWYIQATCATSGDGLYEGLDWLSNQLRNQKSpecifications
ARF1 | |
Polyclonal | |
Mouse polyclonal antibody raised against a partial recombinant ARF1. | |
ARF1 | |
ARF1 | |
ARF1 (NP_001649, 82 a.a. to 181 a.a) partial recombinant protein with GST tag. | |
RUO | |
375 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
ELISA | |
Unconjugated | |
50% glycerol | |
NM_001658 | |
Mouse | |
50 μL | |
Primary | |
Human | |
Antisera |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only