Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARID4A, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89105302 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-302 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-302 Supplier Abnova Corporation Supplier No. H00005926A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ARID4A.

The protein encoded by this gene is a ubiquitously expressed nuclear protein. It binds directly, with several other proteins, to retinoblastoma protein (pRB) which regulates cell proliferation. pRB represses transcription by recruiting the encoded protein. This protein, in turn, serves as a bridging molecule to recruit HDACs and, in addition, provides a second HDAC-independent repression function. The encoded protein possesses transcriptional repression activity. Multiple alternatively spliced transcripts have been observed for this gene, although not all transcript variants have been fully described. [provided by RefSeq

Sequence: AEESQEGLCERESANGFETNVASGTCSIIVQERESREKGQKRPSDGNSGLMAKKQKRTPKRTSAAAKNEKNGTGQSSDSEDLPVLDNSSKCTPVKHLNVSKPQKLAR

Specifications

Antigen ARID4A
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ARID4A.
Formulation 50% glycerol
Gene ARID4A
Gene Accession No. NM_002892
Gene Alias RBBP1/RBP-1/RBP1
Gene Symbols ARID4A
Host Species Mouse
Immunogen ARID4A (NP_002883, 1033 a.a. to 1139 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5926
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.