Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ARSE, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104114 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-114 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-114 Supplier Abnova Corporation Supplier No. H00000415A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ARSE.

Arylsulfatase E is a member of the sulfatase family. It is glycosylated postranslationally and localized to the golgi apparatus. Sulfatases are essential for the correct composition of bone and cartilage matrix. X-linked chondrodysplasia punctata, a disease characterized by abnormalities in cartilage and bone development, has been linked to mutations in this gene. [provided by RefSeq

Sequence: GACYGRKVCPCFGEKVVHHDPPLLFDLSRDPSETHILTPASEPVFYQVMERVQQAVWEHQRTLSPVPLQLDRLGNIWRPWLQPCCGPFPLCWCLREDDPQ

Specifications

Antigen ARSE
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ARSE.
Formulation 50% glycerol
Gene ARSE
Gene Accession No. NM_000047
Gene Alias CDPX/CDPX1/CDPXR/MGC163310
Gene Symbols ARSE
Host Species Mouse
Immunogen ARSE (NP_000038, 490 a.a. to 589 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 415
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.