Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ART4, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104116 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-116 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-116 Supplier Abnova Corporation Supplier No. H00000420A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant ART4.

This gene encodes a protein that contains a mono-ADP-ribosylation (ART) motif. It is a member of the ADP-ribosyltransferase gene family but enzymatic activity has not been demonstrated experimentally. Antigens of the Dombrock blood group system are located on the gene product, which is glycosylphosphatidylinosotol-anchored to the erythrocyte membrane. Allelic variants, some of which lead to adverse transfusion reactions, are known. [provided by RefSeq

Sequence: SFDDQYQGCSKQVMEKLTQGDYFTKDIEAQKNYFRMWQKAHLAWLNQGKVLPQNMTTTHAVAILFYTLNSNVHSDFTRAMASVARTPQQYERSFHFKYLH

Specifications

Antigen ART4
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant ART4.
Formulation 50% glycerol
Gene ART4
Gene Accession No. NM_021071
Gene Alias CD297/DO/DOK1
Gene Symbols ART4
Host Species Mouse
Immunogen ART4 (NP_066549, 61 a.a. to 160 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 420
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.