Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP1A1, Mouse, Clone: 464.6, Abnova™

Catalog No. 89135172 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-172 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-172 Supplier Abnova Corporation Supplier No. MAB2407
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ATP1A1.

Specifications

Antigen ATP1A1
Applications Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 464.6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ATP1A1.
Dilution Flow Cytometry (1:50-1:200) Immunocytochemistry (1:50-1:200) Immunofluorescence (1:100-1:1000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Western Blot (1:1000-1:10000) The optimal working dilution should be
Formulation In ascites (0.09% sodium azide)
Host Species Mouse
Immunogen Native purified rabbit ATP1A1.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100302415
Test Specificity This antibody is specific to Na, K-ATPase alpha 1 subunit.
Target Species Canine, Human, Mouse, Pig, Primate, Rabbit, Rat, Sheep, Xenopus species
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.