Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP1B1, Mouse, Clone: 464.8 (also known as 8A), Abnova™

Catalog No. 89110930 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-930 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-930 Supplier Abnova Corporation Supplier No. MAB2323
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native ATP1B1.

Specifications

Antigen ATP1B1
Applications Western Blot
Classification Monoclonal
Clone 464.8 (also known as
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native ATP1B1.
Dilution Western Blot (1:2500-1:10000) The optimal working dilution should be determined by the end user.
Formulation In buffer containing 0.09% sodium azide
Gene ATP1B1
Gene Symbols ATP1B1
Host Species Mouse
Immunogen Native purified rabbit ATP1B1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100008749
Test Specificity This antibody is specific to Na, K-ATPase beta 1 subunit.
Target Species Bovine, Canine, Human, Pig, Rabbit, Rat
Content And Storage Store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.