Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATP2B4, Mouse, Clone: 2C7, Abnova™

Catalog No. 89104127 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-127 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-127 Supplier Abnova Corporation Supplier No. H00000493M03A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant ATP2B4.

The protein encoded by this gene belongs to the family of P-type primary ion transport ATPases characterized by the formation of an aspartyl phosphate intermediate during the reaction cycle. These enzymes remove bivalent calcium ions from eukaryotic cells against very large concentration gradients and play a critical role in intracellular calcium homeostasis. The mammalian plasma membrane calcium ATPase isoforms are encoded by at least four separate genes and the diversity of these enzymes is further increased by alternative splicing of transcripts. The expression of different isoforms and splice variants is regulated in a developmental, tissue- and cell type-specific manner, suggesting that these pumps are functionally adapted to the physiological needs of particular cells and tissues. This gene encodes the plasma membrane calcium ATPase isoform 4. Alternatively spliced transcript variants encoding different isoforms have been identified. [provided by RefSeq

Sequence: MTNPSDRVLPANSMAESREGDFGCTVMELRKLMELRSRDALTQINVHYGGVQNLCSRLKTSPVEGLSGNPADLEKRRQVFGHNVIPPKKPKT

Specifications

Antigen ATP2B4
Applications ELISA, Western Blot
Classification Cocktail
Clone 2C7
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant ATP2B4.
Formulation ascites with no preservative
Gene ATP2B4
Gene Accession No. NM_001684
Gene Alias ATP2B2/DKFZp686G08106/DKFZp686M088/MXRA1/PMCA4/PMCA4b/PMCA4x
Gene Symbols ATP2B4
Host Species Mouse
Immunogen ATP2B4 (NP_001675,1 a.a. ∽ 92 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 493
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype Ig Mix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.