Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-ATRX, Clone: 23C,

Catalog No. MABE897MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE897MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE897MI Supplier MilliporeSigma™ Supplier No. MABE897
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects ATRX clone: 23C in Human, Mouse samples, and it is validated for Immunocytochemistry, Western Blotting

Transcriptional regulator ATRX (EC 3.6.4.12; UniProt P46100; also known as Alpha thalassemia/mental retardation syndrome X-linked, ATP-dependent helicase ATRX, DNA dependent ATPase and helicase, Helicase 2 X-linked, X-linked helicase II, X-linked nuclear protein, XNP, Zinc finger helicase, Znf-HX) is encoded by the PRF1 (Also known as ATR2, JMS, MRXHF1, RAD54, RAD54L, SFM1, SHS, XH2, XNP, ZNF-HX) gene in human (Gene ID 546). Rad54 belongs to the SWI/SNF family of chromatin remodeling proteins. Rad54 undergoes cell cycle-dependent phosphorylation, nuclear matrix and chromatin association, and is implicatetd in playing a role in gene regulation during interphase and mitosis. ATRX mutations are associated with an X-linked mental retardation (XLMR) syndrome. most often accompanied by alpha-thalassemia syndrome, as a result of diverse changes in DNA methylation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ATRX
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 23C
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. P46100
Gene Symbols ATRX; RAD54L; XH2
Host Species Mouse
Immunogen Recombinant protein corresponding to the N-terminus of human ATRX.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics and Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_000480
Target Species Human, Mouse
Content And Storage Stable for one year at 2°C to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.