Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AVPR1A, Mouse, Clone: 7B8, Abnova™

Catalog No. 89104134 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-134 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-134 Supplier Abnova Corporation Supplier No. H00000552M07A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant AVPR1A.

The protein encoded by this gene acts as receptor for arginine vasopressin. This receptor belongs to the subfamily of G-protein coupled receptors which includes AVPR1B, V2R and OXT receptors. Its activity is mediated by G proteins which stimulate a phosphatidylinositol-calcium second messenger system. The receptor mediates cell contraction and proliferation, platelet aggregation, release of coagulation factor and glycogenolysis. [provided by RefSeq

Sequence: MRLSAGPDAGPSGNSSPWWPLATGAGNTSREAEALGEGNGPPRDVRNEELAK

Specifications

Antigen AVPR1A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 7B8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant AVPR1A.
Formulation ascites with no preservative
Gene AVPR1A
Gene Accession No. NM_000706
Gene Alias AVPR1
Gene Symbols AVPR1A
Host Species Mouse
Immunogen AVPR1A (NP_000697,1 a.a. ∽ 52 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 552
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.