Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Mouse anti-AXL, Clone: 7E10,

Catalog No. MABC680MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC680MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC680MI Supplier MilliporeSigma™ Supplier No. MABC680
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects AXL clone: 7E10 in Human samples, and it is validated for Immunocytochemistry, Western Blotting

AXL receptor tyrosine kinase, also known as UFO, JTK11. AXL is a member of the receptor tyrosine kinase subfamily. Although it is similar to other receptor tyrosine kinases, this protein represents a unique structure of the extracellular region that juxtaposes IgL and FNIII repeats. AXL transduces signals from the extracellular matrix into the cytoplasm by binding growth factors like vitamin K-dependent protein growth-arrest-specific gene 6. AXL is involved in the stimulation of cell proliferation and can also mediate cell aggregation by homophilic binding. Alternatively spliced transcript variants encoding different isoforms have been identified.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AXL
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 7E10
Formulation Mouse monoclonal IgG1 ascitic fluid containing 0.03% sodium azide.
Gene Accession No. P30530
Gene Symbols AXL; UFO
Host Species Mouse
Immunogen Purified recombinant extracellular fragment of human AXL fused with hIgGFc tag expressed in HEK293 cell line.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer
Primary or Secondary Primary
Target Species Human
Content And Storage Stable for one year at -20°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles which may damage IgG and affect product performance.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.