Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azin1, Mouse, Clone: HI-12, Abnova™

Catalog No. 89110597 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-597 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-597 Supplier Abnova Corporation Supplier No. MAB1726

Mouse monoclonal antibody raised against native Azin1.

Specifications

Antigen Azin1
Applications Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone HI-12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native Azin1.
Dilution Western Blot (0.25 ug/mL) Immunocytochemistry (30 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.1% proclin, 2.0% Block Ace)
Gene Azin1
Gene Alias Oazi/Oazin
Gene Symbols Azin1
Host Species Mouse
Immunogen Native purified rat Azin1.
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 58961
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.