Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-cells, Mouse, Clone: 68-IB3, Abnova™

Catalog No. 89135930 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-135-930 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-135-930 Supplier Abnova Corporation Supplier No. MAB5295

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse monoclonal antibody raised against native B-cells.

Specifications

Antigen B-cells
Applications Flow Cytometry, Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 68-IB3
Conjugate Unconjugated
Formulation In PBS (50% glycerol, 0.01% sodium azide)
Host Species Mouse
Immunogen Native purified from WAG/Rij spleen cells.
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The monoclonal antibody recognizes rat B-cells and is reactive with all common inbred strains.
Target Species Human, Rat
Content And Storage Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.