Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B2M, Mouse, Clone: D2A10, Abnova™

Catalog No. 89113360 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
500 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-113-360 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-113-360 Supplier Abnova Corporation Supplier No. MAB8113
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant B2M.

This gene encodes a serum protein found in association with the major histocompatibility complex (MHC) class I heavy chain on the surface of nearly all nucleated cells. The protein has a predominantly beta-pleated sheet structure that can form amyloid fibrils in some pathological conditions. A mutation in this gene has been shown to result in hypercatabolic hypoproteinemia

Specifications

Antigen B2M
Applications ELISA
Classification Monoclonal
Clone D2A10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant B2M.
Dilution The optimal working dilution should be determined by the end user.
Formulation Lyophilized from 10mM PBS
Gene B2M
Gene Symbols B2M
Host Species Mouse
Immunogen Recombinant protein corresponding to human B2M.
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 567
Target Species Human
Content And Storage Store at -20°C.
Aliquot after reconstitution to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.