Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B2M, Mouse, Monoclonal Antibody, Abnova™

Catalog No. 89112248 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-248 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-248 Supplier Abnova Corporation Supplier No. MAB5428

Mouse monoclonal antibody raised against B2M.

This gene encodes a serum protein found in association with the major histocompatibility complex (MHC) class I heavy chain on the surface of nearly all nucleated cells. The protein has a predominantly beta-pleated sheet structure that can form amyloid fibrils in some pathological conditions. A mutation in this gene has been shown to result in hypercatabolic hypoproteinemia

Specifications

Antigen B2M
Applications ELISA, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against B2M.
Dilution ELISA (1:1000000) Western Blot (1:1000-1:2000) The optimal working dilution should be determined by the end user.
Gene B2M
Gene Symbols B2M
Host Species Mouse
Immunogen Human B2M.
Purification Method Protein G affinity chromatography
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 567
Test Specificity This antibody detects specifically target derived from human recombinant protein as well as Conditioned medium from LNCaP cells of at various concentrations. Cross reactivity with mouse and rat are not tested yet.
Target Species Human
Content And Storage Store at -20°C . After reconstitution with sterile PBS store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.