Learn More
B2M, Mouse, PE, Clone: B2M-01, Abnova™
Mouse monoclonal antibody raised against B2M.
Supplier: Abnova Corporation MAB6503
Description
This gene encodes a serum protein found in association with the major histocompatibility complex (MHC) class I heavy chain on the surface of nearly all nucleated cells. The protein has a predominantly beta-pleated sheet structure that can form amyloid fibrils in some pathological conditions. A mutation in this gene has been shown to result in hypercatabolic hypoproteinemia
Specifications
B2M | |
Monoclonal | |
0.1 mg/mL | |
Mouse monoclonal antibody raised against B2M. | |
In PBS (0.2% BSA, 0.09% sodium azide) | |
B2M | |
Purified isolated human B2M. | |
RUO | |
567 | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (PFA fixed), Radioimmune Assays (RIA), Western Blot | |
B2M-01 | |
PE | |
The optimal working dilution should be determined by the end user. | |
B2M | |
Mouse | |
100 μg | |
Primary | |
This antibody reacts with beta2-microglobulin (B2M) associated with cell-surface MHC Class I molecules and other membrane antigens as well as with solublebeta2-microglobulin. | |
Store in the dark at 4°C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing. |
|
IgG2a |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.