Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GAT1, Mouse, Clone: VC1.1, Abnova™

Catalog No. 89111725 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-725 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-725 Supplier Abnova Corporation Supplier No. MAB4196

Mouse monoclonal antibody raised against native B3GAT1.

The protein encoded by this gene is a member of the glucuronyltransferase gene family. These enzymes exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product functions as the key enzyme in a glucuronyl transfer reaction during the biosynthesis of the carbohydrate epitope HNK-1 (human natural killer-1, also known as CD57 and LEU7). Alternate transcriptional splice variants have been characterized. [provided by RefSeq

Specifications

Antigen B3GAT1
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone VC1.1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native B3GAT1.
Dilution Flow Cytometry (1ug stain 1.0x106 cells) The optimal working dilution should be determined by the end user.
Formulation In 10mM PBS, pH 7.2 (1% BSA, 0.09% sodium azide)
Gene B3GAT1
Gene Alias CD57/GLCATP/GlcAT-P/GlcUAT-P/HNK-1/HNK1/LEU7/NK-1
Gene Symbols B3GAT1
Host Species Mouse
Immunogen Native purified B3GAT1 from cat unfixed cortical homogenate area 17.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27087
Test Specificity Recognizes a (110KDa) carbohydrate molecule on myeloid cells. This surface antigen is associated with myelin-associated glycoprotein (MAG). The CD57 antigen is present on 15-20% of normal peripheral blood mononuclear cells. It is expressed on a subset of natural killer cells (60%) and on a subset of T-lymphocytes. This carbohydrate is also present on N-CAM in the nervous system.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.