Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B3GAT3, Mouse, Clone: 4C12, Abnova™

Catalog No. 89106689 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-689 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-689 Supplier Abnova Corporation Supplier No. H00026229M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant B3GAT3.

The protein encoded by this gene is a member of the glucuronyltransferase gene family, enzymes that exhibit strict acceptor specificity, recognizing nonreducing terminal sugars and their anomeric linkages. This gene product catalyzes the formation of the glycosaminoglycan-protein linkage by way of a glucuronyl transfer reaction in the final step of the biosynthesis of the linkage region of proteoglycans. [provided by RefSeq

Sequence: VVGFHTAWEPSRPFPVDMAGFAVALPLLLDKPNAQFDSTAPRGHLESSLLSHLVDPKDLEPRAANCTRVLVWHTRTEKPKMKQEEQLQRQGRGSDPAIEV

Specifications

Antigen B3GAT3
Applications ELISA, Western Blot
Classification Monoclonal
Clone 4C12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant B3GAT3.
Formulation ascites with no preservative
Gene B3GAT3
Gene Accession No. NM_012200
Gene Alias GLCATI/GlcAT-I
Gene Symbols B3GAT3
Host Species Mouse
Immunogen B3GAT3 (NP_036332,236 a.a. ∽ 335 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26229
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.