Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAAT, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89104136 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-136 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-136 Supplier Abnova Corporation Supplier No. H00000570A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant BAAT.

The protein encoded by this gene is a liver enzyme that catalyzes the transfer of C24 bile acids from the acyl-CoA thioester to either glycine or taurine, the second step in the formation of bile acid-amino acid conjugates. The bile acid conjugates then act as a detergent in the gastrointestinal tract, which enhances lipid and fat-soluble vitamin absorption. Defects in this gene are a cause of familial hypercholanemia (FHCA). Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq

Sequence: NGTNFPFGIPQVYHGQIHQPLPHSAQLISTNALGLLELYRTFETTQVGASQYLFPIEEAQGQFLFIVGEGDKTINSKAHAEQAIGQLKRHGKNNWTLL

Specifications

Antigen BAAT
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant BAAT.
Formulation 50% glycerol
Gene BAAT
Gene Accession No. NM_001701
Gene Alias BACAT/BAT/FLJ20300/MGC104432
Gene Symbols BAAT
Host Species Mouse
Immunogen BAAT (NP_001692, 258 a.a. to 355 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 570
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Antisera
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.