Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse anti-Baculovirus Envelope gp64 protein, Clone: AcV1, Novus Biologicals™

Mouse Monoclonal Antibody

$277.00 - $366.50

Specifications

Antigen Baculovirus Envelope gp64 protein
Clone AcV1
Concentration 0.5 mg/mL
Immunogen The immunogen for this antibody is baculovirus envelope gp64 protein.
Formulation Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP20020810
View Documents
Novus Biologicals
NBP200208100UG
100 ug
Each for $277.00
Add to cart
 
NBP20020855
View Documents
Novus Biologicals
NBP200208500UG
500 ug
Each for $366.50
Add to cart
 
Description

Description

Baculovirus Envelope gp64 protein Monoclonal antibody specifically detects Baculovirus Envelope gp64 protein in Virus samples. It is validated for Flow Cytometry.
Specifications

Specifications

Baculovirus Envelope gp64 protein
0.5 mg/mL
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Primary
Mouse
Unconjugated
Protein A or G purified
Flow Cytometry
AcV1
The immunogen for this antibody is baculovirus envelope gp64 protein.
Monoclonal
Store at 4C. Do not freeze.
IgG2a
Affinity Purified
RUO
SDS
Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit