Learn More
BAG3, Mouse, Clone: 5A8, Abnova™
Mouse monoclonal antibody raised against a full-length recombinant BAG3.
Supplier: Abnova Corporation H00009531M01A
Description
BAG proteins compete with Hip for binding to the Hsc70/Hsp70 ATPase domain and promote substrate release. All the BAG proteins have an approximately 45-amino acid BAG domain near the C terminus but differ markedly in their N-terminal regions. The protein encoded by this gene contains a WW domain in the N-terminal region and a BAG domain in the C-terminal region. The BAG domains of BAG1, BAG2, and BAG3 interact specifically with the Hsc70 ATPase domain in vitro and in mammalian cells. All 3 proteins bind with high affinity to the ATPase domain of Hsc70 and inhibit its chaperone activity in a Hip-repressible manner. [provided by RefSeq
Sequence: MSAATHSPMMQVASGNGDRDPLPPGWEIKIDPQTGWPFFVDHNSRTTTWNDPRVPSEGPKETPSSANGPSREGSRLPPAREGHPVYPQLRPGYIPIPVLHEGAENRQVHPFHVYPQPGMQRFRTEAAAAAPQRSQSPLRGMPETTQPDKQCGQVAAAAAAQPPASHGPERSQSPAASDCSSSSSSASLPSSGRSSLGSHQLPRGYISIPVIHEQNVTRPAAQPSFHQAQKTHYPAQQGEYQTHQPVYHKIQGDDWEPRPLRAASPFRSSVQGASSREGSPARSSTPLHSPSPIRVHTVVDRPQQPMTHRETAPVSQPENKPESKPGPVGPELPPGHIPIQVIRKEVDSKPVSQKPPPPSEKVEVKVPPAPVPCPPPSPGPSAVPSSPKSVATEERAAPSTAPAEATPPKPGEAEAPPKHPGVLKVEAILEKVQGLEQAVDNFEGKKTDKKYLMIEEYLTKELLALDSVDPEGRADVRQARRDGVRKVQTILEKLEQKAIDVPGQVQVYELQPSNLEADQPLQAIMEMGAVAADKGKKNAGNAEDPHTETQQPEATAAATSNPSSMTDTPGNPAAPSpecifications
BAG3 | |
Monoclonal | |
Unconjugated | |
ascites with no preservative | |
BC014656 | |
BAG3 | |
BAG3 (AAH14656.1,1 a.a. ∽ 575 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
9531 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Ascites |
ELISA, Western Blot | |
5A8 | |
Mouse monoclonal antibody raised against a full-length recombinant BAG3. | |
BAG3 | |
BAG-3/BIS/CAIR-1/MGC104307 | |
Mouse | |
200 μL | |
Primary | |
Human | |
Antibody | |
IgG2b κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.