Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAG5, Mouse, Clone: 1E3, Abnova™

Catalog No. 89106096 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-106-096 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-106-096 Supplier Abnova Corporation Supplier No. H00009529M02A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant BAG5.

The protein encoded by this gene is a member of the BAG1-related protein family. BAG1 is an anti-apoptotic protein that functions through interactions with a variety of cell apoptosis and growth related proteins including BCL-2, Raf-protein kinase, steroid hormone receptors, growth factor receptors and members of the heat shock protein 70kDa family. This protein contains a BAG domain near the C-terminus, which could bind and inhibit the chaperone activity of Hsc70/Hsp70. Three transcript variants encoding two different isoforms have been found for this gene. [provided by RefSeq

Sequence: MDMGNQHPSISRLQEIQKEVKSVEQQVIGFSGLSDDKNYKKLERILTKQLFEIDSVDTEGKGDIQQARKRAAQETERLLKELEQNANHPHRIEIQNIFEEAQSLVREKIVPFYNGGNCVTDEFEEGIQDIILRLTHVKTGGKISLRKARYHTLTKIWAVQEIIEDCMKKQPSLPLSEDAHPSVAKINFVMCEVNKARGVLIALLMGVNNNETCRHLSCVLSGLIADLDALDVCGRTEIRNYRREVVEDINKLLKYLDLEEEADTTKAFDLRQNHSILKIEKVLKRMREIKNELLQAQNPSELYLSSKTELQGLIGQLDEVSLEKNPCIREARRRAVIEVQTLITYIDLKEALEKRKLFACEEHPSHKAVWNVLGNLSEIQGEVLSFDGNRTDKNYIRLEELLTKQLLALDAVDPQGEEKCKAARKQAVRLAQNILSYLDLKSDEWEY

Specifications

Antigen BAG5
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant BAG5.
Formulation ascites with no preservative
Gene BAG5
Gene Accession No. BC050551
Gene Alias BAG-5
Gene Symbols BAG5
Host Species Mouse
Immunogen BAG5 (AAH50551,1 a.a. ∽ 447 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9529
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.