Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BAK1, Mouse, Clone: 1E4, Abnova™

Catalog No. 89104139 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-104-139 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-104-139 Supplier Abnova Corporation Supplier No. H00000578M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BAK1.

The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form oligomers or heterodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein localizes to mitochondria, and functions to induce apoptosis. It interacts with and accelerates the opening of the mitochondrial voltage-dependent anion channel, which leads to a loss in membrane potential and the release of cytochrome c. This protein also interacts with the tumor suppressor P53 after exposure to cell stress. [provided by RefSeq

Sequence: LQPTAENAYEYFTKIATSLFESGINWGRVVALLGFGYRLALHVYQHGLTGFLGQVTRFVVDFMLHHCIARWIAQRGGWVAALNLGNGPILNVLVVLGVVLLGQFVVRRFFKS

Specifications

Antigen BAK1
Applications ELISA, Immunoprecipitation
Classification Monoclonal
Clone 1E4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BAK1.
Formulation ascites with no preservative
Gene BAK1
Gene Accession No. BC004431
Gene Alias BAK/BAK-LIKE/BCL2L7/CDN1/MGC117255/MGC3887
Gene Symbols BAK1
Host Species Mouse
Immunogen BAK1 (AAH04431,100 a.a. ∽ 211 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 578
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.