Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BARX2, Mouse, Clone: 2B8, Abnova™

Catalog No. 89105880 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-105-880 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-105-880 Supplier Abnova Corporation Supplier No. H00008538M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BARX2.

This gene encodes a member of the homeobox transcription factor family. A highly related protein in mouse has been shown to influence cellular processes that control cell adhesion and remodeling of the actin cytoskeleton in myoblast fusion and chondrogenesis. The encoded protein may also play a role in cancer progression. [provided by RefSeq

Sequence: PDRLDLAQSLGLTQLQVKTWYQNRRMKWKKMVLKGGQEAPTKPKGRPKKNSIPTSEEIEAEEKMNSQAQGQEQLEPSQGQEELCEAQEPKARDVPLEMAE

Specifications

Antigen BARX2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BARX2.
Formulation ascites with no preservative
Gene BARX2
Gene Accession No. NM_003658
Gene Alias MGC133368/MGC133369
Gene Symbols BARX2
Host Species Mouse
Immunogen BARX2 (NP_003649,161 a.a. ∽ 260 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8538
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.