Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BBC3, Mouse, Clone: 2A9G5, Abnova™
Mouse monoclonal antibody raised against recombinant BBC3.
Supplier: Abnova Corporation MAB2755
Specifications
BBC3 | |
Monoclonal | |
Unconjugated | |
Western Blot (2.5-5 ug/mL) The optimal working dilution should be determined by the end user. | |
BBC3 | |
BBC3 | |
Recombinant protein corresponding to amino acids 76-170 of human BBC3. | |
RUO | |
27113 | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Western Blot | |
2A9G5 | |
Mouse monoclonal antibody raised against recombinant BBC3. | |
In PBS (0.02% sodium azide) | |
JFY1/PUMA | |
Mouse | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction