Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAM, Mouse, Clone: BRIC221, Abnova™

Catalog No. 89112186 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-186 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-186 Supplier Abnova Corporation Supplier No. MAB5244
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against native BCAM.

Lutheran blood group glycoprotein is a member of the immunoglobulin superfamily and a receptor for the extracellular matrix protein, laminin. The protein contains five, N-terminus, extracellular immunoglobulin domains, a single transmembrane domain, and a short, C-terminal cytoplasmic tail. This protein may play a role in epithelial cell cancer and in vaso-occlusion of red blood cells in sickle cell disease. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Specifications

Antigen BCAM
Applications Flow Cytometry, Western Blot
Classification Monoclonal
Clone BRIC221
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against native BCAM.
Dilution Flow Cytometry (10ul of the suggested working dilution to label 106 cells or 100ul) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.5 (0.09% sodium azide)
Gene BCAM
Gene Alias AU/CD239/LU/MSK19
Gene Symbols BCAM
Host Species Mouse
Immunogen Native purified BCAM from human erythrocytes.
Quantity 200 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4059
Test Specificity This antibody recognizes a non-polymorphic determinant on both the 85 and 78kD Lutheran (Lu) glycoproteins. BRIC 221 specifically recognizes an epitope in the fourth extracellular domain of Lu glycoprotein. CD239 is expressed by erythrocytes in the peripheral blood.
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.