Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCAP31, Mouse, Clone: A1/182, Abnova™

Catalog No. 89112149 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-149 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-149 Supplier Abnova Corporation Supplier No. MAB5149
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against BCAP31.

Specifications

Antigen BCAP31
Applications ELISA, Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone A1/182
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against BCAP31.
Dilution Western Blot (1:1000) Immunocytochemistry (1:1000) The optimal working dilution should be determined by the end user.
Formulation Lyophilized from purified antibody
Gene BCAP31
Gene Symbols BCAP31
Host Species Mouse
Immunogen Monkey BAP31.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 696663
Test Specificity Recognizes human and monkey BAP31.
Target Species Human, Monkey
Content And Storage Store at -20°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.