Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL10, Mouse, Clone: 4F8E8H8, Abnova™

Catalog No. 89112302 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-112-302 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-112-302 Supplier Abnova Corporation Supplier No. MAB5567
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant BCL10.

Specifications

Antigen BCL10
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Western Blot
Classification Monoclonal
Clone 4F8E8H8
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against recombinant BCL10.
Dilution ELISA (1:10000) Western Blot (1:2000-1:5000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100-1:500) Immunohistochemistry (Frozen sections) (1:200-1:1000) The optimal working dilution should be determined by the end
Formulation In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
Gene BCL10
Gene Alias CARMEN/CIPER/CLAP/c-E10/mE10
Gene Symbols BCL10
Host Species Mouse
Immunogen Recombinant protein corresponding to BCL10.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8915
Target Species Human
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.