Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl2, Mouse, Clone: 10C2, Abnova™

Catalog No. 89111313 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-111-313 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-111-313 Supplier Abnova Corporation Supplier No. MAB3285
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide Bcl2.

Specifications

Antigen Bcl2
Applications ELISA, Western Blot
Classification Monoclonal
Clone 10C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide Bcl2.
Dilution Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In 100mM BBS, pH 8.2
Gene Bcl2
Gene Alias AW986256/Bcl-2/C430015F12Rik/D630044D05Rik/D830018M01Rik
Gene Symbols Bcl2
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 61-76 of mouse Bcl2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12043
Test Specificity Mouse and Rat Bcl-2.
Target Species Mouse, Rat
Content And Storage Store at 4°C.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.