Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL2, Mouse, Clone: 10C4, Abnova™

Catalog No. 89110632 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-632 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-632 Supplier Abnova Corporation Supplier No. MAB1794
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of BCL2.

Specifications

Antigen BCL2
Applications Western Blot
Classification Monoclonal
Clone 10C4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of BCL2.
Dilution Western Blot (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.08% sodium azide)
Gene Bcl2
Gene Alias AW986256/Bcl-2/C430015F12Rik/D630044D05Rik/D830018M01Rik
Gene Symbols Bcl2
Host Species Mouse
Immunogen A synthetic peptide corresponding to amino acids 61-76 of mouse BCL2.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12043
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.