Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL2, Mouse, Clone: 8E12, Abnova™

Catalog No. 89108684 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-108-684 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-108-684 Supplier Abnova Corporation Supplier No. MAB10499
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of BCL2.

This gene encodes an integral outer mitochondrial membrane protein that blocks the apoptotic death of some cells such as lymphocytes. Constitutive expression of BCL2, such as in the case of translocation of BCL2 to Ig heavy chain locus, is thought to be the cause of follicular lymphoma. Two transcript variants, produced by alternate splicing, differ in their C-terminal ends. [provided by RefSeq

Specifications

Antigen BCL2
Applications ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry
Classification Monoclonal
Clone 8E12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of BCL2.
Dilution ELISA (1:10000) Immunohistochemistry (1:200-1:1000) Immunofluorescence (1:200-1:1000) Flow cytometry (1:200-1:400) The optimal working dilution should be determined by the end user.
Formulation In ascites (0.03% sodium azide)
Gene BCL2
Gene Alias Bcl-2
Gene Symbols BCL2
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to human BCL2.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 596
Target Species Human, Mouse
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.