Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BDNF Mouse anti-Human, Clone: 37129, R&D Systems™

Catalog No. MAB848100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB848100 100 μg
MAB848SP 25 μg
MAB848500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB848100 Supplier R&D Systems Supplier No. MAB848100
This item is not returnable. View return policy

Mouse Monoclonal Antibody

BDNF Monoclonal antibody MAB848100 Specifically detects BDNF in Human samples. It is validated for ELISA Capture

Specifications

Antigen BDNF
Applications ELISA
Classification Monoclonal
Clone 37129
Conjugate Unconjugated
Dilution ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. P23560
Gene Alias abrineurin, ANON2, brain-derived neurotrophic factor, BULN2, MGC34632, neurotrophin
Gene Symbols BDNF
Host Species Mouse
Immunogen S. frugiperda insect ovarian cell line Sf 21-derived recombinant human BDNF Arg128-Arg247 Accession # P23560
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 627
Test Specificity Detects human BDNF in ELISAs. In sandwich ELISAs, no cross-reactivity or interference was observed with recombinant human (rh) GDNF, rh beta-NGF, rhNT-3, or rhNT-4.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.