Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BECN1, Mouse, Clone: 1B7, Abnova™
Mouse monoclonal antibody raised against BECN1.
Supplier: Abnova Corporation MAB2367
Specifications
BECN1 | |
Monoclonal | |
Unconjugated | |
In Tris-glycine, 150mM NaCl (0.05% sodium azide) | |
ATG6, VPS30, beclin1 | |
Mouse | |
100 μL | |
Primary | |
Bovine, Chicken, Equine, Human, Mouse, Pig, Primate, Rat | |
Liquid |
Immunohistochemistry, Western Blot | |
1B7 | |
beclin 1, autophagy related | |
BECN1 | |
BECN1 | |
Full length human BECN1. | |
RUO | |
8678 | |
Store at 4°C for short term. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction