Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BECN1, Mouse, Clone: 4H10, Abnova™

Catalog No. 89110965 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-110-965 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-110-965 Supplier Abnova Corporation Supplier No. MAB2368

Mouse monoclonal antibody raised against BECN1.

Specifications

Antigen BECN1
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4H10
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against BECN1.
Dilution Western Blot (2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:100) The optimal working dilution should be determined by the end user.
Formulation In Tris-glycine, 150mM NaCl (0.05% sodium azide)
Gene BECN1
Gene Alias ATG6/VPS30/beclin1
Gene Symbols BECN1
Host Species Mouse
Immunogen Full length human BECN1.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8678
Target Species Bovine, Chicken, Equine, Human, Mouse, Pig, Primate, Rat
Content And Storage Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.